Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037343.1 | NP_001032420.1 | 6 | 425 | ||
NM_001323289.1 | NP_001310218.1 | 4 | 338 | |||
NM_003159.2 | NP_003150.1 | 5 | 429 | |||
GenBank mRNA | AK307444.1 | - | 5 | 426 | ||
AL704691.1 | - | 6 | 464 | |||
AY217744.1 | - | 5 | 397 | |||
HQ171445.1 | - | 4 | 176 | |||
Y15057.1 | - | 5 | 397 |