Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099274.1 | NP_001092744.1 | 3, 4 | 749 | ||
NM_012461.2 | NP_036593.2 | 3, 4 | 749 | |||
XM_005267529.3 | XP_005267586.1 | 2, 3 | 575 | |||
XM_011536642.2 | XP_011534944.1 | 3, 4 | 721 | |||
GenBank mRNA | AF195512.1 | - | 3, 4 | 669 | ||
AK225379.1 | - | 3, 4 | 693 | |||
AK294119.1 | - | 2, 3 | 575 | |||
AK315084.1 | - | 3, 4 | 637 | |||
BC005030.1 | - | 3, 4 | 530 | 3690254 | ||
BC019343.1 | - | 3, 4 | 707 | 3505009 | ||
BM549897.1 | - | 3, 4 | 732 |