Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004562.2 | NP_004553.2 | 2, 3 | 303 | ||
NM_013987.2 | NP_054642.2 | 2, 3 | 303 | |||
XM_011535863.1 | XP_011534165.1 | 2, 3 | 303 | |||
XM_017010908.1 | XP_016866397.1 | 2, 3 | 1217 | |||
GenBank mRNA | AB009973.1 | - | 2, 3 | 270 | ||
AB245403.1 | - | 2, 3 | 261 | |||
AF381284.1 | - | 1, 2 | 169 | |||
AK292590.1 | - | 2, 3 | 264 | |||
BC022014.2 | - | 2, 3 | 267 | 4824892 | ||
DB023187.1 | - | 2, 3 | 303 | |||
EF375726.1 | - | 1, 2 | 169 | |||
GU345838.1 | - | 1, 2 | 186 | |||
GU345839.1 | - | 2, 3 | 186 | |||
GU345840.1 | - | 1, 2 | 186 | |||
GU361466.1 | - | 1, 2 | 186 | |||
GU361467.1 | - | 1, 2 | 186 | |||
GU361469.1 | - | 2, 3 | 283 | |||
GU361470.1 | - | 2, 3 | 284 | |||
KC357595.1 | - | 4, 5 | 381 | |||
KJ534911.1 | - | 1, 2 | 169 | |||
KJ534912.1 | - | 1, 2 | 169 | |||
KU178240.1 | - | 1, 2 | 169 | |||
KU178241.1 | - | 1, 2 | 169 | |||
KU178242.1 | - | 1, 2 | 169 |