Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123066.3 | NP_001116538.2 | 9 | 1784 | ||
NM_001123067.3 | NP_001116539.1 | 6 | 746 | |||
NM_001203251.1 | NP_001190180.1 | 6 | 746 | |||
NM_001203252.1 | NP_001190181.1 | 7 | 833 | |||
NM_005910.5 | NP_005901.2 | 7 | 833 | |||
NM_016834.4 | NP_058518.1 | 5 | 659 | |||
NM_016835.4 | NP_058519.3 | 9 | 1784 | |||
NM_016841.4 | NP_058525.1 | 5 | 659 | |||
XM_005257362.4 | XP_005257419.1 | 9 | 1876 | |||
XM_005257364.4 | XP_005257421.1 | 8 | 1789 | |||
XM_005257365.4 | XP_005257422.1 | 9 | 1876 | |||
XM_005257366.3 | XP_005257423.1 | 7 | 1744 | |||
XM_005257367.4 | XP_005257424.1 | 8 | 1678 | |||
XM_005257368.4 | XP_005257425.1 | 8 | 1678 | |||
XM_005257369.4 | XP_005257426.1 | 8 | 811 | |||
XM_005257370.4 | XP_005257427.1 | 7 | 788 | |||
XM_005257371.4 | XP_005257428.1 | 6 | 637 | |||
GenBank mRNA | AK095802.1 | - | 6 | 486 | ||
AK226139.1 | - | 3 | 5796 | |||
AY526356.1 | - | 1 | 67 | |||
AY730549.1 | - | 5 | 424 | |||
BC000558.2 | - | 5 | 408 | |||
BC098281.1 | - | 4 | 357 | |||
BC099721.1 | - | 4 | 357 | |||
BC101936.1 | - | 4 | 357 | |||
BC114504.1 | - | 4 | 357 | |||
BC114948.1 | - | 4 | 357 | |||
BI552187.1 | - | 6 | 601 | |||
BT006772.1 | - | 4 | 337 | |||
J03778.1 | - | 5 | 374 | |||
X14474.1 | - | 4 | 367 |