Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAPT SIRNA Assays ›
Gene Symbol:
MAPT
Gene Name:
microtubule associated protein tau
Gene Aliases:
DDPAC, FTDP-17, MAPTL, MSTD, MTBT1, MTBT2, PPND, PPP1R103, TAU
Chromosome Location:
Chr.17: 45894382 - 46028334 on Build GRCh38
UniGene ID:
Hs.101174
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s530989
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAPT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001123066.3
NP_001116538.2
11
2116
NM_001123067.3
NP_001116539.1
7
1024
NM_001203251.1
NP_001190180.1
7
1024
NM_001203252.1
NP_001190181.1
8
1111
NM_005910.5
NP_005901.2
8
1111
NM_016834.4
NP_058518.1
6
937
NM_016835.4
NP_058519.3
10
2062
NM_016841.4
NP_058525.1
6
937
XM_005257362.4
XP_005257419.1
10
2154
XM_005257364.4
XP_005257421.1
9
2067
XM_005257365.4
XP_005257422.1
10
2154
XM_005257366.3
XP_005257423.1
8
2022
XM_005257367.4
XP_005257424.1
9
1956
XM_005257368.4
XP_005257425.1
9
1956
XM_005257369.4
XP_005257426.1
9
1089
XM_005257370.4
XP_005257427.1
8
1066
XM_005257371.4
XP_005257428.1
7
915
GenBank mRNA
AK095802.1
-
7
764
AK226139.1
-
4
6074
AK299658.1
-
3
354
AK299704.1
-
6
626
AY526356.1
-
2
345
AY730549.1
-
6
702
BC000558.2
-
6
686
BC098281.1
-
5
635
BC099721.1
-
5
635
BC101936.1
-
5
635
BC114504.1
-
5
635
BC114948.1
-
5
635
BT006772.1
-
5
615
J03778.1
-
6
652
X14474.1
-
5
645
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
neuron migration
brain development
female pregnancy
response to nutrient
adult walking behavior
axo-dendritic transport
intrinsic apoptotic signaling pathway in response to oxidative stress
response to organic substance
regulation of autophagy
regulation of microtubule polymerization
positive regulation of microtubule polymerization
negative regulation of intracellular transport
positive regulation of axon extension
mitochondrion transport along microtubule
axon extension
generation of neurons
regulation of microtubule-based movement
structural constituent of cytoskeleton
protein binding
microtubule binding
SH3 domain binding
enzyme binding
protein kinase binding
protein complex binding
apolipoprotein binding
protein phosphatase 2A binding
lipoprotein particle binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline