Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGMO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004320.1 | NP_001004320.1 | 1 | 227 | ||
XM_006715730.1 | XP_006715793.1 | 1 | 227 | |||
XM_006715731.3 | XP_006715794.1 | 1 | 227 | |||
XM_011515402.2 | XP_011513704.1 | 1 | 227 | |||
XM_017012204.1 | XP_016867693.1 | 1 | 227 | |||
GenBank mRNA | AK131284.1 | - | 1 | 227 | ||
BC108676.1 | - | 1 | 174 | 6152531 | ||
KJ534765.1 | - | 1 | 57 |