Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079866.1 | NP_001073335.1 | 8 | 1267 | ||
NM_001257342.1 | NP_001244271.1 | 9 | 1397 | |||
NM_001257343.1 | NP_001244272.1 | 9 | 1449 | |||
NM_001257344.1 | NP_001244273.1 | 8 | 1324 | |||
NM_001318836.1 | NP_001305765.1 | 7 | 898 | |||
NM_001320717.1 | NP_001307646.1 | 9 | 1439 | |||
NM_004328.4 | NP_004319.1 | 9 | 1476 | |||
XM_005246748.2 | XP_005246805.1 | 8 | 1751 | |||
XM_005246749.3 | XP_005246806.1 | 6 | 750 | |||
XM_006712678.1 | XP_006712741.1 | 8 | 1882 | |||
XM_017004631.1 | XP_016860120.1 | 9 | 1638 | |||
XM_017004632.1 | XP_016860121.1 | 9 | 1363 | |||
XM_017004633.1 | XP_016860122.1 | 8 | 2141 | |||
XM_017004634.1 | XP_016860123.1 | 7 | 1169 | |||
GenBank mRNA | AF026849.1 | - | 8 | 1243 | ||
AF038195.1 | - | 8 | 1205 | |||
AK096210.1 | - | 8 | 1830 | |||
AK223518.1 | - | 8 | 1353 | |||
AK289363.1 | - | 9 | 1339 | |||
BC000416.2 | - | 8 | 1397 | 2961496 | ||
BC007500.2 | - | 8 | 1397 | 2961496 | ||
BX571752.1 | - | 9 | 1396 | |||
KP992846.1 | - | 9 | 1396 | |||
KU177941.1 | - | 7 | 1130 | |||
KU177942.1 | - | 6 | 985 |