Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
PLEC
Gene Name:
plectin
Gene Aliases:
EBS1, EBSMD, EBSND, EBSO, EBSOG, EBSPA, HD1, LGMD2Q, PCN, PLEC1, PLEC1b, PLTN
Chromosome Location:
Chr.8: 143915147 - 143976800 on Build GRCh38
UniGene ID:
Hs.434248
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s531207
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000445.4
NP_000436.2
33
13166
NM_201378.3
NP_958780.1
32
13051
NM_201379.2
NP_958781.1
32
13119
NM_201380.3
NP_958782.1
32
13617
NM_201381.2
NP_958783.1
32
13014
NM_201382.3
NP_958784.1
32
13104
NM_201383.2
NP_958785.1
32
13057
NM_201384.2
NP_958786.1
32
13165
XM_005250976.3
XP_005251033.1
33
13659
XM_005250978.3
XP_005251035.1
33
13184
XM_005250979.4
XP_005251036.1
33
13143
XM_005250980.4
XP_005251037.1
33
13228
XM_005250981.3
XP_005251038.1
33
13065
XM_005250982.3
XP_005251039.1
33
13058
XM_005250983.2
XP_005251040.1
32
12982
XM_005250984.4
XP_005251041.1
33
13047
XM_006716588.3
XP_006716651.1
34
13236
XM_006716589.2
XP_006716652.1
33
12997
XM_006716590.3
XP_006716653.1
33
13107
XM_011517130.2
XP_011515432.1
33
13172
XM_011517131.2
XP_011515433.1
32
13091
XM_011517132.2
XP_011515434.1
32
9799
GenBank mRNA
AY480044.1
-
32
13123
AY480045.1
-
32
12994
AY480046.1
-
32
13119
AY480047.1
-
32
13617
AY480048.1
-
32
13014
AY480049.1
-
32
13043
AY480050.1
-
32
13057
AY480051.1
-
32
13165
BC007597.1
-
1
2482
BC013206.1
-
1
431
U53204.1
-
33
13168
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
hemidesmosome assembly
cell-cell adhesion
actin binding
protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0