Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s531543
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
5
492
NM_001018017.2
NP_001018017.1
5
465
NM_001204285.1
NP_001191214.1
5
1125
NM_001204286.1
NP_001191215.1
5
1152
NM_001204287.1
NP_001191216.1
5
546
NM_001204288.1
NP_001191217.1
5
492
NM_001204289.1
NP_001191218.1
4
414
NM_001204290.1
NP_001191219.1
4
351
NM_001204291.1
NP_001191220.1
4
423
NM_001204292.1
NP_001191221.1
4
417
NM_001204293.1
NP_001191222.1
5
519
NM_001204294.1
NP_001191223.1
4
390
NM_002456.5
NP_002447.4
5
519
GenBank mRNA
AF125525.1
-
5
384
AK301575.1
-
6
1377
AY327587.1
-
5
470
AY327588.1
-
4
365
AY327596.1
-
6
600
BC120974.2
-
5
442
40115473
BC120975.2
-
5
469
40115475
EF583642.1
-
5
795
EF583643.1
-
5
623
EF583644.1
-
5
609
EF583645.1
-
5
569
EF583646.1
-
5
537
EF583647.1
-
5
524
EF583648.1
-
5
451
EF583649.1
-
5
470
EF583650.1
-
5
407
EF583651.1
-
5
402
EF583653.1
-
4
350
EF583654.1
-
4
329
EF670706.1
-
5
819
EF670707.1
-
5
488
EF670708.1
-
4
401
EF670709.1
-
4
395
EF670710.1
-
4
387
EF670711.1
-
5
497
EF670712.1
-
4
368
EF670714.1
-
4
323
FJ226028.1
-
4
926
FJ226029.1
-
4
1084
FJ226030.1
-
5
1054
FJ226031.1
-
4
1012
FJ226032.1
-
4
945
FJ226033.1
-
4
926
FJ226034.1
-
5
845
FJ226037.1
-
5
623
FJ226039.1
-
5
550
FJ226040.1
-
5
470
FJ226041.1
-
6
627
FJ226042.1
-
5
524
FJ226043.1
-
5
510
FJ226044.1
-
4
494
FJ226045.1
-
4
464
FJ226047.1
-
5
424
FJ226052.1
-
5
443
J05581.1
-
5
1125
J05582.1
-
3466
M34088.1
-
3
612
M34089.1
-
3
612
U60259.1
-
4
343
U60260.1
-
5
418
U60261.1
-
5
472
X52228.1
-
4
1479
X52229.1
-
6
1230
X80761.1
-
5
393
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-dvnl5:80/10.239.162.83:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0