Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s531544
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
8
920
NM_001018017.2
NP_001018017.1
8
893
NM_001044390.2
NP_001037855.1
7
737
NM_001044391.2
NP_001037856.1
5
578
NM_001044392.2
NP_001037857.1
5
605
NM_001044393.2
NP_001037858.1
7
705
NM_001204285.1
NP_001191214.1
8
1553
NM_001204286.1
NP_001191215.1
8
1580
NM_001204287.1
NP_001191216.1
8
974
NM_001204288.1
NP_001191217.1
8
888
NM_001204289.1
NP_001191218.1
7
842
NM_001204290.1
NP_001191219.1
7
779
NM_001204291.1
NP_001191220.1
7
851
NM_001204292.1
NP_001191221.1
7
845
NM_001204293.1
NP_001191222.1
7
825
NM_001204294.1
NP_001191223.1
7
818
NM_001204295.1
NP_001191224.1
6
695
NM_001204296.1
NP_001191225.1
7
764
NM_001204297.1
NP_001191226.1
5
659
NM_002456.5
NP_002447.4
8
947
GenBank mRNA
AK301575.1
-
9
1805
AY327587.1
-
8
898
AY327588.1
-
7
793
AY327596.1
-
9
1028
BC120974.2
-
8
870
40115473
BC120975.2
-
8
897
40115475
EF583642.1
-
8
1223
EF583643.1
-
8
1051
EF583644.1
-
8
1037
EF583646.1
-
8
965
EF583647.1
-
8
952
EF583648.1
-
8
879
EF583649.1
-
8
866
EF583650.1
-
8
835
EF583651.1
-
8
830
EF583652.1
-
7
820
EF583653.1
-
7
778
EF583654.1
-
7
757
EF583655.1
-
6
705
EF583656.1
-
5
566
EF670706.1
-
8
1247
EF670707.1
-
8
916
EF670708.1
-
7
829
EF670709.1
-
7
823
EF670710.1
-
7
815
EF670711.1
-
7
803
EF670712.1
-
7
796
EF670713.1
-
7
761
EF670714.1
-
7
751
EF670715.1
-
5
745
EF670716.1
-
6
673
EF670717.1
-
6
658
EF670718.1
-
5
637
EF670719.1
-
5
607
EF670720.1
-
4
567
EF670721.1
-
5
564
EF670722.1
-
5
541
EF670723.1
-
3
337
FJ226028.1
-
8
1516
FJ226029.1
-
7
1512
FJ226030.1
-
8
1482
FJ226031.1
-
7
1440
FJ226032.1
-
7
1373
FJ226033.1
-
7
1354
FJ226034.1
-
8
1273
FJ226035.1
-
7
1256
FJ226036.1
-
6
1236
FJ226037.1
-
9
1213
FJ226038.1
-
5
1179
FJ226039.1
-
9
1140
FJ226040.1
-
9
1060
FJ226041.1
-
9
1055
FJ226042.1
-
9
1027
FJ226043.1
-
8
938
FJ226044.1
-
7
922
FJ226045.1
-
7
892
FJ226046.1
-
7
887
FJ226047.1
-
8
852
FJ226049.1
-
7
815
FJ226050.1
-
7
788
FJ226051.1
-
7
761
FJ226052.1
-
7
749
FJ226053.1
-
7
742
FJ226054.1
-
6
738
FJ226055.1
-
6
686
J05581.1
-
8
1553
J05582.1
-
3892
M34088.1
-
6
1040
M34089.1
-
6
1040
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0