Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDPCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042692.2 | NP_001036157.1 | 3 | 456 | ||
NM_015910.5 | NP_056994.3 | 9 | 1271 | |||
NR_122106.1 | 3 | 456 | ||||
XM_005264348.3 | XP_005264405.1 | 9 | 1311 | |||
XM_011532881.2 | XP_011531183.1 | 10 | 1095 | |||
XM_011532884.2 | XP_011531186.1 | 9 | 1311 | |||
XM_011532886.2 | XP_011531188.1 | 9 | 1311 | |||
XM_011532887.2 | XP_011531189.1 | 9 | 1311 | |||
XM_011532890.2 | XP_011531192.1 | 9 | 1311 | |||
XM_011532891.2 | XP_011531193.1 | 10 | 1296 | |||
XM_017004251.1 | XP_016859740.1 | 11 | 1949 | |||
XM_017004252.1 | XP_016859741.1 | 10 | 1095 | |||
XM_017004253.1 | XP_016859742.1 | 9 | 1311 | |||
XM_017004254.1 | XP_016859743.1 | 9 | 1311 | |||
GenBank mRNA | AF131737.1 | - | 3 | 456 | ||
AK001849.1 | - | 1 | 171 | |||
AK055030.1 | - | 10 | 1296 | |||
AK309028.1 | - | 4 | 511 | |||
AK310290.1 | - | 9 | 938 | |||
AL833623.1 | - | 9 | 1057 | |||
BC093752.1 | - | 3 | 401 | 7939597 | ||
BC093754.1 | - | 3 | 401 | 7939599 | ||
BX538331.1 | - | 9 | 1017 | |||
BX647132.1 | - | 3 | 3784 |