Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005373.2 | NP_005364.1 | 6 | 924 | ||
XM_017001320.1 | XP_016856809.1 | 5 | 2024 | |||
GenBank mRNA | DQ234353.1 | - | 6 | 858 | ||
M90102.1 | - | 6 | 879 | |||
M90103.1 | - | 6 | 879 |