Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000029.3 | NP_000020.1 | 2 | 791 | ||
GenBank mRNA | AB209769.1 | - | 2 | 303 | ||
AK222798.1 | - | 2 | 322 | |||
AK222815.1 | - | 2 | 323 | |||
AK293507.1 | - | 2 | 322 | |||
AK303755.1 | - | 3 | 264 | |||
AK307978.1 | - | 3 | 237 | |||
AK312291.1 | - | 2 | 401 | |||
AU128714.1 | - | 2 | 497 | |||
BC011519.1 | - | 2 | 311 | 4213559 | ||
BF338744.1 | - | 2 | 394 | |||
BT006851.1 | - | 1 | 283 | |||
K02215.1 | - | 2 | 322 | |||
M69110.1 | - | 1 | 297 |