Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: FKBP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021939.3 | NP_068758.3 | 1 | 540 | ||
XM_011525099.2 | XP_011523401.1 | 1 | 988 | |||
XM_011525100.2 | XP_011523402.1 | 1 | 261 | |||
GenBank mRNA | AB045981.1 | - | 1 | 301 | ||
AF337909.1 | - | 1 | 215 | |||
AK092042.1 | - | 1 | 261 | |||
AK092708.1 | - | 1 | 540 | |||
AK172811.1 | - | 1 | 246 | |||
AK296107.1 | - | 1 | 253 | |||
AL832985.1 | - | 1 | 282 | |||
BC016467.1 | - | 1 | 275 | 4154344 | ||
BX537452.1 | - | 1 | 295 |