Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256534.1 | NP_001243463.1 | 8 | 1166 | ||
NM_001287387.1 | NP_001274316.1 | 9 | 1020 | |||
NM_005984.4 | NP_005975.1 | 9 | 1185 | |||
NR_046298.2 | 8 | 1046 | ||||
GenBank mRNA | AK298204.1 | - | 9 | 1018 | ||
BC018590.1 | - | 9 | 943 | |||
L77567.1 | - | 9 | 1067 | |||
U25147.1 | - | 9 | 1069 |