Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM53AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028346.1 | 2, 3 | 612 | |||
GenBank mRNA | JF968456.1 | - | 1, 2 | 135 | ||
JF968457.1 | - | 1, 2 | 135 |
Gene Symbol: TRIM49D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206627.1 | NP_001193556.1 | 1, 2 | 417 | ||
XM_011542816.2 | XP_011541118.1 | 2, 3 | 533 |
Gene Symbol: TRIM49D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105522.1 | NP_001098992.1 | 2, 3 | 531 |
Gene Symbol: TRIM53CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JF968458.1 | - | 1, 2 | 135 |