Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM49D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105522.1 | NP_001098992.1 | 7 | 1207 |
Gene Symbol: TRIM49D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206627.1 | NP_001193556.1 | 6 | 1093 | ||
XM_011542816.2 | XP_011541118.1 | 7 | 1209 | |||
GenBank mRNA | BX116917.1 | - | 1 | 113 |