Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DST SIRNA Assays ›
Gene Symbol:
DST
Gene Name:
dystonin
Gene Aliases:
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location:
Chr.6: 56457987 - 56954671 on Build GRCh38
UniGene ID:
Hs.604915
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s532081
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144769.2
NP_001138241.1
20
2724
NM_001144770.1
NP_001138242.1
18
2395
NM_001723.5
NP_001714.1
7
1279
NM_015548.4
NP_056363.2
7
1279
NM_183380.3
NP_899236.1
17
2669
XM_005249310.3
XP_005249367.1
21
2818
XM_005249315.3
XP_005249372.1
20
2765
XM_005249316.2
XP_005249373.1
13
3041
XM_005249318.2
XP_005249375.1
19
2479
XM_005249319.2
XP_005249376.1
18
2380
XM_005249320.4
XP_005249377.1
18
2436
XM_005249322.4
XP_005249379.1
18
2331
XM_005249323.3
XP_005249380.1
17
2664
XM_005249324.2
XP_005249381.1
7
1276
XM_011514824.2
XP_011513126.1
20
2729
XM_011514825.2
XP_011513127.1
20
2725
XM_011514826.2
XP_011513128.1
17
2664
XM_017011205.1
XP_016866694.1
21
2828
XM_017011206.1
XP_016866695.1
21
2828
XM_017011207.1
XP_016866696.1
21
2765
XM_017011208.1
XP_016866697.1
21
2828
XM_017011209.1
XP_016866698.1
21
2828
XM_017011210.1
XP_016866699.1
21
2828
XM_017011211.1
XP_016866700.1
21
2828
XM_017011212.1
XP_016866701.1
21
2828
XM_017011213.1
XP_016866702.1
21
2828
XM_017011214.1
XP_016866703.1
21
2828
XM_017011215.1
XP_016866704.1
21
2828
XM_017011216.1
XP_016866705.1
21
2828
XM_017011217.1
XP_016866706.1
18
2409
XM_017011219.1
XP_016866708.1
21
2824
XM_017011220.1
XP_016866709.1
20
2767
XM_017011221.1
XP_016866710.1
21
2824
XM_017011222.1
XP_016866711.1
13
2983
XM_017011223.1
XP_016866712.1
21
2824
XM_017011224.1
XP_016866713.1
17
2664
XM_017011225.1
XP_016866714.1
21
2838
GenBank mRNA
AF400226.1
-
7
1171
AF400227.1
-
7
1171
AK294830.1
-
17
2277
AK295864.1
-
13
2651
AK299898.1
-
18
2395
BC065536.1
-
7
1243
L11690.1
-
7
1273
M69225.1
-
7
1209
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
cell adhesion
integrin-mediated signaling pathway
response to wounding
maintenance of cell polarity
intermediate filament cytoskeleton organization
cell motility
actin binding
integrin binding
calcium ion binding
protein binding
protein C-terminus binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-7lstp:80/10.239.162.105:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0