Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000577.4 | NP_000568.1 | 3 | 265 | ||
NM_001318914.1 | NP_001305843.1 | 5 | 499 | |||
NM_173841.2 | NP_776213.1 | 4 | 328 | |||
NM_173842.2 | NP_776214.1 | 2 | 257 | |||
NM_173843.2 | NP_776215.1 | 4 | 436 | |||
XM_005263661.4 | XP_005263718.1 | 3 | 284 | |||
XM_011511121.1 | XP_011509423.1 | 7 | 787 | |||
GenBank mRNA | AF043143.1 | - | 4 | 436 | ||
AK290898.1 | - | 4 | 318 | |||
AK290926.1 | - | 2 | 259 | |||
BC009745.2 | - | 3 | 170 | 3689284 | ||
BI766516.1 | - | 2 | 257 | |||
DA667440.1 | - | 5 | 416 | |||
M55646.1 | - | 3 | 261 | |||
X52015.1 | - | 2 | 216 | |||
X53296.1 | - | 2 | 201 | |||
X84348.1 | - | 4 | 234 |