Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC52A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033409.3 | NP_212134.3 | 5 | 2181 | ||
XM_005260655.3 | XP_005260712.1 | 6 | 2110 | |||
XM_011529148.1 | XP_011527450.1 | 6 | 2619 | |||
GenBank mRNA | AK074650.1 | - | 4 | 2542 | ||
BC009750.2 | - | 5 | 2084 | 3689286 |
Set Membership: |
![]() |