Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020312.3 | NP_064708.1 | 4 | 556 | ||
GenBank mRNA | AF161444.1 | - | 4 | 490 | ||
AK290627.1 | - | 4 | 486 | |||
AK293585.1 | - | 4 | 490 | |||
AK295793.1 | - | 4 | 499 | |||
AK303527.1 | - | 4 | 479 | |||
BC001478.2 | - | 2 | 131 | |||
BC029341.1 | - | 4 | 490 | |||
BC054340.1 | - | 4 | 490 | 6514554 | ||
BC064946.1 | - | 4 | 491 | 6023461 |