Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUTYH SIRNA Assays ›
Gene Symbol:
MUTYH
Gene Name:
mutY DNA glycosylase
Gene Aliases:
MYH
Chromosome Location:
Chr.1: 45329242 - 45340925 on Build GRCh38
UniGene ID:
Hs.271353
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s532602
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUTYH
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001048171.1
NP_001041636.1
14
1616
NM_001048172.1
NP_001041637.1
14
1507
NM_001048173.1
NP_001041638.1
14
1504
NM_001048174.1
NP_001041639.1
14
1423
NM_001128425.1
NP_001121897.1
14
1658
NM_001293190.1
NP_001280119.1
14
1619
NM_001293191.1
NP_001280120.1
14
1456
NM_001293192.1
NP_001280121.1
14
1552
NM_001293195.1
NP_001280124.1
15
1573
NM_001293196.1
NP_001280125.1
14
1441
NM_012222.2
NP_036354.1
14
1649
XM_011541497.2
XP_011539799.1
14
1715
XM_011541498.1
XP_011539800.1
14
1493
XM_011541499.1
XP_011539801.1
14
1479
XM_011541500.2
XP_011539802.1
14
1674
XM_011541501.2
XP_011539803.1
15
1588
XM_011541502.2
XP_011539804.1
14
1493
XM_011541503.2
XP_011539805.1
13
1461
XM_011541504.2
XP_011539806.1
14
1542
XM_011541505.2
XP_011539807.1
10
1162
XM_011541506.1
XP_011539808.1
10
1146
XM_011541507.2
XP_011539809.2
9
958
XM_017001331.1
XP_016856820.1
14
1492
XM_017001332.1
XP_016856821.1
14
1494
XM_017001333.1
XP_016856822.1
14
1547
XM_017001334.1
XP_016856823.1
14
1440
XM_017001335.1
XP_016856824.1
14
1386
XM_017001336.1
XP_016856825.1
13
1470
XM_017001337.1
XP_016856826.1
13
1410
XM_017001338.1
XP_016856827.1
13
1997
XM_017001339.1
XP_016856828.1
13
1448
GenBank mRNA
AB032920.1
-
14
1567
AB032921.1
-
14
1537
AB032922.1
-
14
1534
AB032923.1
-
14
1470
AB032924.1
-
14
1434
AB032925.1
-
14
1401
AB032926.1
-
15
1551
AB032927.1
-
14
1472
AB032928.1
-
14
1469
AB032929.1
-
14
1405
AK293830.1
-
5
678
BC003178.1
-
14
1554
2958002
KU178203.1
-
14
1400
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA glycosylase
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
mismatch repair
response to oxidative stress
depurination
DNA binding
protein binding
DNA N-glycosylase activity
MutLalpha complex binding
MutLbeta complex binding
MutSalpha complex binding
MutSbeta complex binding
metal ion binding
4 iron, 4 sulfur cluster binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline