Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PORCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203473.2 | NP_982299.1 | 6, 7 | 865 | ||
NM_203475.2 | NP_982301.1 | 6, 7 | 865 | |||
XM_005272635.2 | XP_005272692.1 | 6, 7 | 1220 | |||
XM_005272637.1 | XP_005272694.1 | 7, 8 | 1147 | |||
XM_006724545.3 | XP_006724608.1 | 7, 8 | 930 | |||
XM_006724546.3 | XP_006724609.1 | 7, 8 | 1002 | |||
XM_006724547.1 | XP_006724610.1 | 6, 7 | 722 | |||
XM_017029735.1 | XP_016885224.1 | 6, 7 | 1230 | |||
GenBank mRNA | AF317058.1 | - | 5, 6 | 686 | ||
AF317059.1 | - | 5, 6 | 686 | |||
AK314745.1 | - | 5, 6 | 735 | |||
BC019080.2 | - | 6, 7 | 812 | 4559533 | ||
L08239.1 | - | 4, 5 | 602 |