Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOLH1 SIRNA Assays ›
Gene Symbol:
FOLH1
FOLH1B
Gene Name:
folate hydrolase (prostate-specific membrane antigen) 1
folate hydrolase 1B
Gene Aliases:
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
FOLH2, FOLHP, GCP3, GCPIII, PSM, PSMA-LIKE, PSMAL
Chromosome Location:
Chr.11: 49145094 - 49208706 on Build GRCh38
Chromosome Location:
Chr.11: 89637575 - 89698718 on Build GRCh38
UniGene ID:
Hs.645352
Hs.645352
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s5328
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FOLH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014986.1
NP_001014986.1
2
414
NM_001193471.1
NP_001180400.1
3
511
NM_001193472.1
NP_001180401.1
3
511
NM_001193473.1
NP_001180402.1
2
414
NM_004476.1
NP_004467.1
2
414
XM_011519958.2
XP_011518260.2
3
548
XM_017017432.1
XP_016872921.1
3
546
XM_017017433.1
XP_016872922.1
3
546
XM_017017434.1
XP_016872923.1
3
212
XM_017017435.1
XP_016872924.1
3
211
XM_017017436.1
XP_016872925.1
2
446
XM_017017437.1
XP_016872926.1
2
173
XM_017017438.1
XP_016872927.1
2
446
XM_017017439.1
XP_016872928.1
3
247
XM_017017440.1
XP_016872929.1
2
117
XM_017017441.1
XP_016872930.1
2
58
XM_017017442.1
XP_016872931.1
2
184
XM_017017443.1
XP_016872932.1
2
68
XM_017017444.1
XP_016872933.1
2
348
XM_017017445.1
XP_016872934.1
3
475
XM_017017446.1
XP_016872935.1
2
446
XM_017017447.1
XP_016872936.1
2
116
XM_017017448.1
XP_016872937.1
2
108
XM_017017449.1
XP_016872938.1
2
348
XM_017017450.1
XP_016872939.1
3
475
XM_017017451.1
XP_016872940.1
2
117
GenBank mRNA
AF176574.1
-
2
291
AJ007318.1
-
3
168
AK295368.1
-
2
346
AK295470.1
-
3
443
AK300532.1
-
2
346
AK312366.1
-
2
342
AY101595.1
-
2
153
BC025672.1
-
2
312
5202715
DQ088979.1
-
2
276
EF488811.2
-
3
213
M99487.1
-
2
414
S76978.1
-
2
146
Gene Symbol: FOLH1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017017319.1
XP_016872808.1
2
108
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
folic acid-containing compound metabolic process
cellular amino acid biosynthetic process
C-terminal protein deglutamylation
metallocarboxypeptidase activity
peptidase activity
dipeptidase activity
metal ion binding
Ac-Asp-Glu binding
tetrahydrofolyl-poly(glutamate) polymer binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0