Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133468.4 | NP_597725.1 | 5 | 716 | ||
XM_005249633.2 | XP_005249690.1 | 4 | 456 | |||
XM_017011800.1 | XP_016867289.1 | 4 | 376 | |||
XM_017011801.1 | XP_016867290.1 | 4 | 376 | |||
GenBank mRNA | AK289963.1 | - | 4 | 456 | ||
AY324650.1 | - | 5 | 715 | |||
BC060868.1 | - | 4 | 393 | 30345290 |