Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CHAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142929.1 | NP_001136401.1 | 7 | 1255 | ||
NM_001142933.1 | NP_001136405.1 | 8 | 1325 | |||
NM_001142934.1 | NP_001136406.1 | 8 | 1325 | |||
NM_020549.4 | NP_065574.3 | 7 | 1255 | |||
NM_020984.3 | NP_066264.3 | 7 | 921 | |||
NM_020985.3 | NP_066265.3 | 8 | 1083 | |||
NM_020986.3 | NP_066266.3 | 7 | 911 | |||
GenBank mRNA | AF305907.1 | - | 7 | 1255 | ||
AF305908.2 | - | 8 | 1325 | |||
AF305909.1 | - | 7 | 921 | |||
BC130615.1 | - | 6 | 781 | 40146646 | ||
BC130617.1 | - | 6 | 781 | 40146648 | ||
S45018.1 | - | 6 | 772 | |||
S56138.1 | - | 7 | 1255 |