Gene Symbol: PARN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134477.2 | NP_001127949.1 | 12, 13 | 911 | ||
NM_001242992.1 | NP_001229921.1 | 11, 12 | 856 | |||
NM_002582.3 | NP_002573.1 | 12, 13 | 994 | |||
XM_011522510.2 | XP_011520812.1 | 12, 13 | 1000 | |||
XM_011522511.2 | XP_011520813.1 | 12, 13 | 999 | |||
XM_011522513.2 | XP_011520815.1 | 12, 13 | 910 | |||
XM_011522514.1 | XP_011520816.1 | 12, 13 | 998 | |||
XM_017023259.1 | XP_016878748.1 | 9, 10 | 643 | |||
XM_017023260.1 | XP_016878749.1 | 7, 8 | 532 | |||
GenBank mRNA | AJ005698.1 | - | 12, 13 | 905 | ||
AK023866.1 | - | 5, 6 | 279 | |||
AK293189.1 | - | 12, 13 | 903 | |||
AK299653.1 | - | 5, 6 | 436 | |||
AK301648.1 | - | 11, 12 | 846 | |||
AK315020.1 | - | 12, 13 | 990 | |||
BC044763.1 | - | 12, 13 | 932 | |||
BC050029.1 | - | 12, 13 | 932 | 4825218 |