Gene Symbol: PARN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134477.2 | NP_001127949.1 | 16 | 1117 | ||
NM_001242992.1 | NP_001229921.1 | 15 | 1062 | |||
NM_002582.3 | NP_002573.1 | 16 | 1200 | |||
XM_011522510.2 | XP_011520812.1 | 16 | 1206 | |||
XM_011522511.2 | XP_011520813.1 | 16 | 1205 | |||
XM_011522513.2 | XP_011520815.1 | 16 | 1116 | |||
XM_011522514.1 | XP_011520816.1 | 16 | 1204 | |||
XM_017023258.1 | XP_016878747.1 | 15 | 1126 | |||
XM_017023259.1 | XP_016878748.1 | 12 | 805 | |||
XM_017023260.1 | XP_016878749.1 | 10 | 694 | |||
GenBank mRNA | AJ005698.1 | - | 16 | 1111 | ||
AK023866.1 | - | 9 | 485 | |||
AK293189.1 | - | 16 | 1109 | |||
AK299653.1 | - | 9 | 642 | |||
AK301648.1 | - | 15 | 1052 | |||
AK315020.1 | - | 16 | 1196 | |||
BC044763.1 | - | 16 | 1138 | |||
BC050029.1 | - | 16 | 1138 | 4825218 |