Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYBPC1 SIRNA Assays ›
Gene Symbol:
MYBPC1
Gene Name:
myosin binding protein C, slow type
Gene Aliases:
LCCS4, MYBPCC, MYBPCS
Chromosome Location:
Chr.12: 101594849 - 101695841 on Build GRCh38
UniGene ID:
Hs.654589
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533124
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYBPC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001254718.1
NP_001241647.1
14
1575
NM_001254719.1
NP_001241648.1
14
1575
NM_001254720.1
NP_001241649.1
13
1539
NM_001254721.1
NP_001241650.1
13
1518
NM_001254722.1
NP_001241651.1
12
1497
NM_001254723.1
NP_001241652.1
13
1536
NM_002465.3
NP_002456.2
16
1650
NM_206819.2
NP_996555.1
16
1650
NM_206820.2
NP_996556.1
14
1575
NM_206821.2
NP_996557.1
14
1575
XM_005268876.4
XP_005268933.1
15
1584
XM_006719405.3
XP_006719468.1
16
1511
XM_006719406.3
XP_006719469.1
16
1511
XM_006719407.3
XP_006719470.1
15
1584
XM_006719408.3
XP_006719471.1
16
1511
XM_006719409.3
XP_006719472.1
14
1537
XM_006719410.3
XP_006719473.1
16
1511
XM_006719411.3
XP_006719474.1
14
1537
XM_017019315.1
XP_016874804.1
15
1454
XM_017019316.1
XP_016874805.1
16
1511
XM_017019317.1
XP_016874806.1
14
1537
XM_017019318.1
XP_016874807.1
15
1454
XM_017019319.1
XP_016874808.1
13
1480
XM_017019320.1
XP_016874809.1
12
1579
XM_017019321.1
XP_016874810.1
13
1480
XM_017019322.1
XP_016874811.1
11
1522
GenBank mRNA
AK292976.1
-
16
1610
AK295727.1
-
12
1457
AK296681.1
-
14
1539
AK303401.1
-
13
1496
AL831993.2
-
16
1585
AL832000.2
-
16
1585
AL832332.2
-
16
1585
AL832354.2
-
16
1587
AL832456.2
-
16
1585
AL834249.2
-
16
1585
BC015187.1
-
16
1610
BC092418.1
-
14
1522
30529254
BC117217.1
-
12
1432
40125768
BC143495.1
-
13
1453
9052003
BC143502.1
-
12
1432
9052010
BC143503.1
-
14
1510
9052011
BC143504.1
-
13
1474
9052012
BX649073.2
-
17
1681
X66276.1
-
14
1525
X73114.1
-
14
1516
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
muscle filament sliding
positive regulation of protein targeting to mitochondrion
actin binding
protein binding
structural constituent of muscle
titin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f4tdd:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0