Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYBPC1 SIRNA Assays ›
Gene Symbol:
MYBPC1
Gene Name:
myosin binding protein C, slow type
Gene Aliases:
LCCS4, MYBPCC, MYBPCS
Chromosome Location:
Chr.12: 101594849 - 101695841 on Build GRCh38
UniGene ID:
Hs.654589
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533125
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYBPC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001254718.1
NP_001241647.1
28
3528
NM_001254719.1
NP_001241648.1
27
3474
NM_001254720.1
NP_001241649.1
26
3438
NM_001254721.1
NP_001241650.1
26
3417
NM_001254722.1
NP_001241651.1
25
3396
NM_001254723.1
NP_001241652.1
26
3435
NM_002465.3
NP_002456.2
29
3549
NM_206819.2
NP_996555.1
29
3549
NM_206820.2
NP_996556.1
28
3528
NM_206821.2
NP_996557.1
27
3474
XM_005268876.4
XP_005268933.1
28
3483
XM_006719405.3
XP_006719468.1
30
3464
XM_006719406.3
XP_006719469.1
30
3464
XM_006719407.3
XP_006719470.1
29
3537
XM_006719408.3
XP_006719471.1
29
3410
XM_006719409.3
XP_006719472.1
28
3490
XM_006719410.3
XP_006719473.1
30
3464
XM_006719411.3
XP_006719474.1
28
3490
XM_017019315.1
XP_016874804.1
29
3407
XM_017019316.1
XP_016874805.1
30
3464
XM_017019317.1
XP_016874806.1
27
3436
XM_017019318.1
XP_016874807.1
28
3353
XM_017019319.1
XP_016874808.1
27
3433
XM_017019320.1
XP_016874809.1
26
3532
XM_017019321.1
XP_016874810.1
26
3379
XM_017019322.1
XP_016874811.1
24
3421
GenBank mRNA
AK056587.1
-
16
1657
AK057284.1
-
9
1593
AK292976.1
-
29
3509
AK295727.1
-
25
3356
AK296681.1
-
28
3492
AK303401.1
-
26
3395
AL831993.2
-
29
3484
AL832000.2
-
29
3484
AL832332.2
-
29
3484
AL832350.2
-
29
3484
AL832456.2
-
29
3484
AL834249.2
-
29
3484
BC092418.1
-
27
3421
30529254
BC117217.1
-
25
3331
40125768
BC143495.1
-
26
3352
9052003
BC143502.1
-
25
3331
9052010
BC143503.1
-
27
3409
9052011
BC143504.1
-
26
3373
9052012
BX649073.2
-
30
3579
X66276.1
-
28
3475
X73114.1
-
27
3415
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
muscle filament sliding
positive regulation of protein targeting to mitochondrion
actin binding
protein binding
structural constituent of muscle
titin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0