Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030311.2 | NP_001025482.1 | 2 | 356 | ||
NM_001030312.2 | NP_001025483.1 | 2 | 356 | |||
NM_001030313.2 | NP_001025484.1 | 2 | 356 | |||
NM_001160277.1 | NP_001153749.1 | 2 | 356 | |||
NM_201548.4 | NP_963842.1 | 2 | 356 | |||
NR_027689.1 | 2 | 356 | ||||
NR_027690.1 | 2 | 356 | ||||
GenBank mRNA | AJ640141.1 | - | 2 | 273 | ||
AJ697855.1 | - | 2 | 255 | |||
AJ697856.1 | - | 2 | 255 | |||
AJ697858.1 | - | 2 | 273 | |||
AK293844.1 | - | 2 | 357 | |||
AY357073.1 | - | 2 | 255 | |||
AY690329.1 | - | 2 | 255 | |||
AY690330.1 | - | 2 | 255 | |||
AY690331.1 | - | 2 | 255 | |||
AY690332.1 | - | 2 | 255 | |||
AY690333.1 | - | 2 | 255 | |||
BC137498.1 | - | 2 | 282 | 9021498 | ||
BC137499.1 | - | 2 | 282 | 9021499 | ||
HQ426666.1 | - | 2 | 162 | |||
HQ426667.1 | - | 2 | 162 | |||
HQ426668.1 | - | 2 | 162 | |||
HQ426669.1 | - | 2 | 162 | |||
HQ426670.1 | - | 2 | 162 | |||
HQ426671.1 | - | 2 | 84 | |||
HQ426672.1 | - | 2 | 84 | |||
HQ426673.1 | - | 2 | 84 | |||
HQ426674.1 | - | 2 | 84 | |||
HQ426675.1 | - | 2 | 84 | |||
HQ426676.1 | - | 2 | 84 | |||
HQ426678.1 | - | 3 | 409 | |||
HQ426679.1 | - | 1 | 17 | |||
JF290420.1 | - | 2 | 242 |