Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533223
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
41
6163
NM_182961.3
NP_892006.3
41
6307
XM_006715407.1
XP_006715470.1
41
6205
XM_006715408.2
XP_006715471.1
41
6209
XM_006715409.1
XP_006715472.1
40
6184
XM_006715410.2
XP_006715473.1
41
6209
XM_006715411.1
XP_006715474.1
40
6154
XM_006715412.2
XP_006715475.1
41
6209
XM_006715413.2
XP_006715476.1
41
6209
XM_006715414.1
XP_006715477.1
39
6133
XM_006715415.2
XP_006715478.1
41
6209
XM_006715416.2
XP_006715479.1
41
6209
XM_006715417.2
XP_006715480.1
41
6209
XM_006715420.2
XP_006715483.1
41
6209
XM_006715421.2
XP_006715484.1
41
6209
XM_006715422.1
XP_006715485.1
40
6046
XM_006715423.2
XP_006715486.1
41
6209
XM_006715424.2
XP_006715487.1
41
6209
XM_006715425.2
XP_006715488.1
41
6209
XM_011535641.2
XP_011533943.1
41
6209
XM_011535642.2
XP_011533944.1
41
6209
XM_011535643.1
XP_011533945.1
40
6040
XM_011535644.1
XP_011533946.1
27
4256
XM_011535645.2
XP_011533947.1
23
3649
XM_017010608.1
XP_016866097.1
42
6324
XM_017010609.1
XP_016866098.1
41
6131
XM_017010610.1
XP_016866099.1
40
6110
XM_017010611.1
XP_016866100.1
40
5913
XM_017010612.1
XP_016866101.1
39
5837
XM_017010613.1
XP_016866102.1
41
6209
XM_017010614.1
XP_016866103.1
41
6209
XM_017010615.1
XP_016866104.1
41
6209
XM_017010616.1
XP_016866105.1
41
6209
XM_017010617.1
XP_016866106.1
41
6209
XM_017010618.1
XP_016866107.1
41
6209
XM_017010619.1
XP_016866108.1
27
4288
GenBank mRNA
AF495910.1
-
41
6307
AF535142.1
-
41
6163
FM162565.1
-
39
5923
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f4tdd:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0