Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533225
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
105
19845
NM_182961.3
NP_892006.3
106
20223
XM_006715407.1
XP_006715470.1
106
20136
XM_006715408.2
XP_006715471.1
106
20128
XM_006715409.1
XP_006715472.1
105
20115
XM_006715410.2
XP_006715473.1
106
20140
XM_006715411.1
XP_006715474.1
105
20085
XM_006715412.2
XP_006715475.1
106
20125
XM_006715413.2
XP_006715476.1
106
20140
XM_006715414.1
XP_006715477.1
104
20064
XM_006715415.2
XP_006715478.1
106
20140
XM_006715416.2
XP_006715479.1
106
20125
XM_006715417.2
XP_006715480.1
105
19999
XM_006715420.2
XP_006715483.1
105
19987
XM_006715421.2
XP_006715484.1
105
19984
XM_006715422.1
XP_006715485.1
105
19977
XM_006715423.2
XP_006715486.1
106
20140
XM_006715424.2
XP_006715487.1
106
20140
XM_006715425.2
XP_006715488.1
106
20140
XM_011535641.2
XP_011533943.1
106
20137
XM_011535642.2
XP_011533944.1
106
20125
XM_011535643.1
XP_011533945.1
105
19971
XM_011535644.1
XP_011533946.1
92
18187
XM_011535645.2
XP_011533947.1
88
17580
XM_017010608.1
XP_016866097.1
107
20255
XM_017010609.1
XP_016866098.1
106
20062
XM_017010610.1
XP_016866099.1
105
20041
XM_017010611.1
XP_016866100.1
105
19844
XM_017010612.1
XP_016866101.1
104
19768
XM_017010613.1
XP_016866102.1
106
20137
XM_017010614.1
XP_016866103.1
105
19984
XM_017010615.1
XP_016866104.1
105
19984
XM_017010616.1
XP_016866105.1
106
20140
XM_017010617.1
XP_016866106.1
106
20137
XM_017010618.1
XP_016866107.1
106
20125
XM_017010619.1
XP_016866108.1
92
18219
GenBank mRNA
AF495910.1
-
106
20223
AF535142.1
-
105
19845
AY061755.1
-
21
3313
AY184206.1
-
19
3208
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cnw2s:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0