Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161683.1 | NP_001155155.1 | 5 | 664 | ||
NM_144672.3 | NP_653273.3 | 9 | 808 | |||
XM_011545747.2 | XP_011544049.1 | 10 | 1187 | |||
XM_017022950.1 | XP_016878439.1 | 10 | 911 | |||
GenBank mRNA | AK125840.1 | - | 5 | 664 | ||
BC129992.1 | - | 9 | 805 | |||
BC129993.1 | - | 9 | 812 | 40128985 |