Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127891.2 | NP_001121363.1 | 6 | 1146 | ||
NM_001302508.1 | NP_001289437.1 | 6 | 960 | |||
NM_001302509.1 | NP_001289438.1 | 6 | 889 | |||
NM_001302510.1 | NP_001289439.1 | 6 | 882 | |||
NM_004530.5 | NP_004521.1 | 6 | 1288 | |||
GenBank mRNA | AK301536.1 | - | 6 | 889 | ||
AK310314.1 | - | 6 | 1146 | |||
AK312711.1 | - | 6 | 1254 | |||
AL832088.1 | - | 7 | 1435 | |||
AU279927.1 | - | 2 | 185 | |||
BC002576.2 | - | 6 | 1243 | 3161383 | ||
DQ385623.1 | - | 1 | 85 | |||
J03210.1 | - | 6 | 930 |