Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533401
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
7, 8
1118
NM_001243226.2
NP_001230155.2
8, 9
908
NM_001243227.1
NP_001230156.1
6, 7
952
NM_001243228.1
NP_001230157.1
7, 8
1044
NM_001243230.1
NP_001230159.1
6, 7
594
NM_001243231.1
NP_001230160.1
5, 6
552
NM_001243232.1
NP_001230161.1
3, 4
574
NM_001243233.1
NP_001230162.1
3, 4
515
NM_001306207.1
NP_001293136.1
6, 7
743
NM_001306208.1
NP_001293137.1
3, 4
574
NM_003199.2
NP_003190.1
7, 8
1118
XM_005266739.3
XP_005266796.2
6, 7
758
XM_005266741.3
XP_005266798.1
7, 8
718
XM_005266743.4
XP_005266800.1
6, 7
579
XM_005266744.3
XP_005266801.1
6, 7
699
XM_005266745.3
XP_005266802.1
6, 7
561
XM_005266747.3
XP_005266804.1
5, 6
555
XM_005266749.3
XP_005266806.1
3, 4
346
XM_005266752.4
XP_005266809.1
3, 4
916
XM_005266754.3
XP_005266811.1
2, 3
340
XM_005266755.4
XP_005266812.1
3, 4
375
XM_006722536.2
XP_006722599.1
7, 8
1041
XM_006722537.2
XP_006722600.1
7, 8
674
XM_006722538.2
XP_006722601.1
6, 7
696
XM_011526158.2
XP_011524460.1
6, 7
1672
XM_011526160.2
XP_011524462.1
2, 3
725
XM_017025934.1
XP_016881423.1
6, 7
755
XM_017025935.1
XP_016881424.1
6, 7
758
XM_017025936.1
XP_016881425.1
6, 7
758
XM_017025937.1
XP_016881426.1
6, 7
512
XM_017025938.1
XP_016881427.1
7, 8
671
XM_017025939.1
XP_016881428.1
7, 8
721
XM_017025940.1
XP_016881429.1
7, 8
1041
XM_017025941.1
XP_016881430.1
7, 8
674
XM_017025942.1
XP_016881431.1
7, 8
718
XM_017025943.1
XP_016881432.1
7, 8
721
XM_017025944.1
XP_016881433.1
6, 7
976
XM_017025945.1
XP_016881434.1
6, 7
979
XM_017025946.1
XP_016881435.1
6, 7
993
XM_017025947.1
XP_016881436.1
6, 7
579
XM_017025948.1
XP_016881437.1
6, 7
976
XM_017025949.1
XP_016881438.1
6, 7
696
XM_017025950.1
XP_016881439.1
6, 7
976
XM_017025951.1
XP_016881440.1
5, 6
555
XM_017025952.1
XP_016881441.1
3, 4
569
XM_017025953.1
XP_016881442.1
3, 4
346
XM_017025954.1
XP_016881443.1
3, 4
930
XM_017025955.1
XP_016881444.1
2, 3
664
XM_017025956.1
XP_016881445.1
2, 3
340
GenBank mRNA
AB209741.1
-
2, 3
226
AK095041.1
-
3, 4
574
AK096862.1
-
5, 6
552
AK122765.1
-
7, 8
1118
AK299169.1
-
6, 7
952
AK300612.1
-
3, 4
515
AK300636.1
-
2, 3
245
AK301144.1
-
6, 7
729
AK315074.1
-
7, 8
1032
AK316165.1
-
2, 3
289
BC031056.1
-
8, 9
869
BC125084.1
-
6, 7
532
40117795
BC125085.1
-
6, 7
532
40117798
FR748208.1
-
2, 3
161
FR748209.1
-
2, 3
662
FR748210.1
-
6, 7
506
FR748211.1
-
6, 7
506
FR748214.1
-
5, 6
434
FR748216.1
-
6, 7
500
FR748217.1
-
5, 6
380
FR748218.1
-
1, 2
116
FR748219.1
-
3, 4
293
M74719.1
-
7, 8
705
X52079.1
-
4, 5
371
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: AK133885, AK028748, AK081012, X91753, U16322, and others.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-nsq4p:80/10.239.163.53:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0