Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533403
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
14, 15
1752
NM_001243226.2
NP_001230155.2
15, 16
1542
NM_001243227.1
NP_001230156.1
13, 14
1586
NM_001243228.1
NP_001230157.1
14, 15
1696
NM_001243230.1
NP_001230159.1
13, 14
1225
NM_001243231.1
NP_001230160.1
12, 13
1186
NM_001243232.1
NP_001230161.1
10, 11
1208
NM_001243233.1
NP_001230162.1
10, 11
1149
NM_001243234.1
NP_001230163.1
7, 8
781
NM_001243235.1
NP_001230164.1
7, 8
781
NM_001243236.1
NP_001230165.1
7, 8
797
NM_001306207.1
NP_001293136.1
13, 14
1377
NM_001306208.1
NP_001293137.1
10, 11
1208
NM_003199.2
NP_003190.1
14, 15
1752
XM_005266739.3
XP_005266796.2
13, 14
1392
XM_005266741.3
XP_005266798.1
14, 15
1352
XM_005266743.4
XP_005266800.1
13, 14
1213
XM_005266744.3
XP_005266801.1
13, 14
1333
XM_005266745.3
XP_005266802.1
13, 14
1195
XM_005266747.3
XP_005266804.1
12, 13
1189
XM_005266749.3
XP_005266806.1
10, 11
980
XM_005266752.4
XP_005266809.1
10, 11
1550
XM_005266754.3
XP_005266811.1
9, 10
974
XM_005266755.4
XP_005266812.1
10, 11
1009
XM_005266761.3
XP_005266818.1
7, 8
1290
XM_006722536.2
XP_006722599.1
14, 15
1675
XM_006722537.2
XP_006722600.1
14, 15
1308
XM_006722538.2
XP_006722601.1
13, 14
1330
XM_011526158.2
XP_011524460.1
13, 14
2306
XM_011526160.2
XP_011524462.1
9, 10
1359
XM_017025934.1
XP_016881423.1
13, 14
1389
XM_017025935.1
XP_016881424.1
13, 14
1389
XM_017025936.1
XP_016881425.1
13, 14
1392
XM_017025937.1
XP_016881426.1
13, 14
1146
XM_017025938.1
XP_016881427.1
14, 15
1305
XM_017025939.1
XP_016881428.1
14, 15
1352
XM_017025940.1
XP_016881429.1
14, 15
1675
XM_017025941.1
XP_016881430.1
14, 15
1308
XM_017025942.1
XP_016881431.1
14, 15
1352
XM_017025943.1
XP_016881432.1
14, 15
1352
XM_017025944.1
XP_016881433.1
13, 14
1610
XM_017025945.1
XP_016881434.1
13, 14
1610
XM_017025946.1
XP_016881435.1
13, 14
1627
XM_017025947.1
XP_016881436.1
13, 14
1213
XM_017025948.1
XP_016881437.1
13, 14
1610
XM_017025949.1
XP_016881438.1
13, 14
1330
XM_017025950.1
XP_016881439.1
13, 14
1607
XM_017025951.1
XP_016881440.1
12, 13
1186
XM_017025952.1
XP_016881441.1
10, 11
1200
XM_017025953.1
XP_016881442.1
10, 11
980
XM_017025954.1
XP_016881443.1
10, 11
1561
XM_017025955.1
XP_016881444.1
9, 10
1295
XM_017025956.1
XP_016881445.1
9, 10
974
XM_017025957.1
XP_016881446.1
7, 8
1290
GenBank mRNA
AB209741.1
-
9, 10
860
AK095041.1
-
10, 11
1208
AK096862.1
-
12, 13
1186
AK122765.1
-
14, 15
1752
AK299169.1
-
13, 14
1586
AK300038.1
-
7, 8
727
AK300612.1
-
10, 11
1149
AK300636.1
-
9, 10
879
AK301144.1
-
13, 14
1363
AK315074.1
-
15, 16
1684
AK316165.1
-
9, 10
923
AL518903.3
-
7, 8
797
BC031056.1
-
15, 16
1503
BC125084.1
-
13, 14
1166
40117795
BC125085.1
-
13, 14
1166
40117798
FR748210.1
-
13, 14
1140
FR748211.1
-
13, 14
1140
FR748212.1
-
11, 12
960
FR748213.1
-
11, 12
960
FR748214.1
-
12, 13
1068
FR748215.1
-
10, 11
888
FR748216.1
-
13, 14
1131
FR748217.1
-
12, 13
1014
FR748218.1
-
8, 9
750
FR748219.1
-
10, 11
927
FR748220.1
-
7, 8
660
FR748221.1
-
7, 8
660
FR748222.1
-
7, 8
660
FR748223.1
-
5, 6
492
M74718.1
-
7, 8
612
M74719.1
-
14, 15
1339
X52079.1
-
11, 12
1005
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-nsq4p:80/10.239.163.53:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0