Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SETD5 SIRNA Assays ›
Gene Symbol:
SETD5
Gene Name:
SET domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 9397700 - 9478154 on Build GRCh38
UniGene ID:
Hs.288164
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s533414
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SETD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080517.2
NP_001073986.1
17
2813
NM_001292043.1
NP_001278972.1
19
3077
XM_005265301.1
XP_005265358.1
17
2851
XM_005265303.1
XP_005265360.1
19
3232
XM_011533920.1
XP_011532222.1
20
3327
XM_011533921.1
XP_011532223.1
19
2929
XM_011533922.1
XP_011532224.1
18
2890
XM_011533927.1
XP_011532229.1
20
3270
XM_011533928.1
XP_011532230.1
18
2833
XM_011533930.1
XP_011532232.1
20
3271
XM_011533932.1
XP_011532234.1
18
3038
XM_011533933.1
XP_011532235.1
16
2407
XM_017006767.1
XP_016862256.1
20
4989
XM_017006768.1
XP_016862257.1
18
4503
XM_017006769.1
XP_016862258.1
18
6606
XM_017006770.1
XP_016862259.1
19
6645
XM_017006771.1
XP_016862260.1
17
2851
XM_017006772.1
XP_016862261.1
18
6606
XM_017006773.1
XP_016862262.1
18
6549
XM_017006774.1
XP_016862263.1
17
2794
XM_017006775.1
XP_016862264.1
18
6549
XM_017006776.1
XP_016862265.1
19
3134
XM_017006777.1
XP_016862266.1
19
3077
XM_017006778.1
XP_016862267.1
20
3118
XM_017006779.1
XP_016862268.1
18
3095
XM_017006780.1
XP_016862269.1
19
3079
XM_017006781.1
XP_016862270.1
20
3118
XM_017006782.1
XP_016862271.1
19
6645
XM_017006783.1
XP_016862272.1
21
3343
XM_017006784.1
XP_016862273.1
19
6645
XM_017006785.1
XP_016862274.1
20
6758
XM_017006786.1
XP_016862275.1
20
6709
GenBank mRNA
AB051544.1
-
13
2607
AK001569.1
-
6
996
AK024384.1
-
5
907
AK091393.1
-
14
2116
AK092046.1
-
19
3032
AK127298.1
-
7
1700
AK289529.1
-
8
1395
AL442073.1
-
4
793
BC020956.1
-
6
1027
3851178
BX648380.1
-
9
1415
CR627408.1
-
16
2429
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-9xb8z:80/10.239.165.207:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0