Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000411.6 | NP_000402.3 | 7 | 1833 | ||
NM_001242784.1 | NP_001229713.1 | 7 | 2767 | |||
NM_001242785.1 | NP_001229714.1 | 7 | 1926 | |||
XM_005260953.3 | XP_005261010.1 | 6 | 1795 | |||
XM_005260955.3 | XP_005261012.1 | 6 | 1666 | |||
XM_005260956.3 | XP_005261013.1 | 6 | 1842 | |||
XM_006723994.2 | XP_006724057.1 | 6 | 1780 | |||
XM_006723995.1 | XP_006724058.1 | 7 | 1958 | |||
XM_011529538.1 | XP_011527840.1 | 7 | 1813 | |||
XM_011529539.2 | XP_011527841.1 | 7 | 1818 | |||
XM_011529540.2 | XP_011527842.1 | 6 | 1795 | |||
XM_011529541.2 | XP_011527843.1 | 6 | 1669 | |||
XM_017028330.1 | XP_016883819.1 | 6 | 1616 | |||
GenBank mRNA | AK314189.1 | - | 6 | 1700 | ||
BC060787.1 | - | 7 | 1791 | 30343493 | ||
D23672.2 | - | 7 | 1824 | |||
D87328.1 | - | 7 | 2584 | |||
X80160.1 | - | 6 | 1684 |