Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000411.6 | NP_000402.3 | 2 | 189 | ||
NM_001242784.1 | NP_001229713.1 | 2 | 314 | |||
NM_001242785.1 | NP_001229714.1 | 2 | 282 | |||
XM_005260953.3 | XP_005261010.1 | 2 | 297 | |||
XM_005260955.3 | XP_005261012.1 | 2 | 168 | |||
XM_005260956.3 | XP_005261013.1 | 2 | 344 | |||
XM_006723994.2 | XP_006724057.1 | 2 | 282 | |||
XM_006723995.1 | XP_006724058.1 | 2 | 314 | |||
XM_011529538.1 | XP_011527840.1 | 2 | 169 | |||
XM_011529539.2 | XP_011527841.1 | 2 | 198 | |||
XM_011529540.2 | XP_011527842.1 | 2 | 297 | |||
XM_011529541.2 | XP_011527843.1 | 2 | 171 | |||
XM_017028330.1 | XP_016883819.1 | 2 | 118 | |||
GenBank mRNA | AJ001864.1 | - | 2 | 358 | ||
AK307940.1 | - | 2 | 296 | |||
AK314189.1 | - | 1 | 55 | |||
BC060787.1 | - | 2 | 147 | 30343493 | ||
D23672.2 | - | 2 | 180 | |||
D87328.1 | - | 2 | 131 | |||
DA334377.1 | - | 2 | 189 | |||
DB290499.1 | - | 2 | 314 | |||
X80160.1 | - | 1 | 40 |