Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134.2 | NP_001125.1 | 8 | 929 | ||
GenBank mRNA | AK297669.1 | - | 8 | 841 | ||
AK297674.1 | - | 8 | 782 | |||
AK314817.1 | - | 8 | 923 | |||
BC027881.1 | - | 8 | 915 | 5208299 | ||
V01514.1 | - | 8 | 929 |