Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290020.1 | NP_001276949.1 | 15 | 2319 | ||
NM_001290021.1 | NP_001276950.1 | 8 | 1145 | |||
NM_001312689.1 | NP_001299618.1 | 10 | 1536 | |||
NM_018076.4 | NP_060546.2 | 15 | 2272 | |||
XM_011519527.1 | XP_011517829.1 | 15 | 2319 | |||
XM_011519528.1 | XP_011517830.1 | 15 | 2969 | |||
XM_011519533.2 | XP_011517835.1 | 10 | 1493 | |||
XM_011519534.2 | XP_011517836.1 | 10 | 1626 | |||
XM_011519535.2 | XP_011517837.1 | 9 | 1378 | |||
XM_011519537.2 | XP_011517839.1 | 6 | 1053 | |||
XM_017016368.1 | XP_016871857.1 | 15 | 2319 | |||
XM_017016369.1 | XP_016871858.1 | 15 | 2319 | |||
XM_017016370.1 | XP_016871859.1 | 14 | 2109 | |||
GenBank mRNA | AK001679.1 | - | 15 | 2272 | ||
AK292521.1 | - | 15 | 2275 | |||
AK302048.1 | - | 8 | 1099 | |||
AK302419.1 | - | 10 | 1868 | |||
AK302627.1 | - | 10 | 1536 | |||
AL136859.1 | - | 4 | 658 | |||
BC140846.1 | - | 14 | 2200 | 9021714 |