Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143905.2 | NP_001137377.1 | 2 | 357 | ||
NM_001194995.1 | NP_001181924.1 | 2 | 650 | |||
NM_152269.4 | NP_689482.1 | 2 | 441 | |||
XM_005253630.4 | XP_005253687.1 | 3 | 461 | |||
XM_011538980.2 | XP_011537282.1 | 2 | 1219 | |||
GenBank mRNA | AF061733.1 | - | 2 | 283 | ||
AK025908.1 | - | 2 | 280 | |||
AK095982.1 | - | 2 | 821 | |||
AK126043.1 | - | 2 | 4212 | |||
BC018145.1 | - | 2 | 242 | 3846678 | ||
BC020885.1 | - | 2 | 301 | 4665283 | ||
BC062329.1 | - | 2 | 284 | 5484911 | ||
BI850864.1 | - | 2 | 357 | |||
BU507560.1 | - | 2 | 464 |