Gene Symbol: DTNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271667.1 | NP_001258596.1 | 7, 8 | 745 | ||
NM_001271668.1 | NP_001258597.1 | 6, 7 | 649 | |||
NM_001271669.1 | NP_001258598.1 | 5, 6 | 595 | |||
NM_032122.4 | NP_115498.2 | 7, 8 | 700 | |||
NM_183040.2 | NP_898861.1 | 7, 8 | 700 | |||
NR_036448.1 | 8, 9 | 839 | ||||
XM_005249447.4 | XP_005249504.1 | 8, 9 | 701 | |||
XM_011514936.2 | XP_011513238.1 | 7, 8 | 691 | |||
GenBank mRNA | AF061734.1 | - | 3, 4 | 187 | ||
AF394226.1 | - | 7, 8 | 617 | |||
AK054593.1 | - | 7, 8 | 669 | |||
AK290718.1 | - | 7, 8 | 617 | |||
AK310590.1 | - | 8, 9 | 701 | |||
AL136637.2 | - | 7, 8 | 617 | |||
AY265460.1 | - | 7, 8 | 624 | |||
BC011912.2 | - | 7, 8 | 615 | 4139934 | ||
BE793298.1 | - | 557 | ||||
BQ056746.1 | - | 6, 7 | 588 | |||
BX394617.2 | - | 8, 9 | 839 | |||
KU178801.1 | - | 7, 8 | 511 | |||
KU178802.1 | - | 5, 6 | 406 |