Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271667.1 | NP_001258596.1 | 10 | 1091 | ||
NM_001271668.1 | NP_001258597.1 | 9 | 995 | |||
NM_001271669.1 | NP_001258598.1 | 8 | 941 | |||
NM_032122.4 | NP_115498.2 | 10 | 1046 | |||
XM_005249447.4 | XP_005249504.1 | 11 | 1047 | |||
XM_011514936.2 | XP_011513238.1 | 10 | 1037 | |||
XM_011514937.2 | XP_011513239.1 | 4 | 809 | |||
XM_017011348.1 | XP_016866837.1 | 4 | 515 | |||
XM_017011349.1 | XP_016866838.1 | 4 | 436 | |||
GenBank mRNA | AF061734.1 | - | 6 | 533 | ||
AF394226.1 | - | 10 | 963 | |||
AK054593.1 | - | 10 | 1015 | |||
AY265460.1 | - | 10 | 970 | |||
BC011912.2 | - | 10 | 961 | 4139934 | ||
KU178801.1 | - | 10 | 857 | |||
KU178802.1 | - | 8 | 752 |