Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000893.3 | NP_000884.1 | 6 | 933 | ||
NM_001102416.2 | NP_001095886.1 | 6 | 933 | |||
NM_001166451.1 | NP_001159923.1 | 5 | 825 | |||
GenBank mRNA | AK223589.1 | - | 6 | 882 | ||
AK290839.1 | - | 6 | 832 | |||
AK298440.1 | - | 7 | 847 | |||
AK303768.1 | - | 6 | 913 | |||
AK315230.1 | - | 6 | 885 | |||
BC026253.1 | - | 6 | 611 | |||
BC060039.1 | - | 6 | 836 | 4593077 | ||
CD014130.1 | - | 5 | 787 | |||
K02566.1 | - | 6 | 748 |