Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025009.4 | NP_079285.2 | 21 | 3076 | ||
XM_005265788.3 | XP_005265845.1 | 21 | 3043 | |||
XM_006714055.2 | XP_006714118.1 | 21 | 3043 | |||
XM_011534412.2 | XP_011532714.1 | 11 | 1395 | |||
GenBank mRNA | AB014535.1 | - | 14 | 2729 | ||
BC136535.1 | - | 21 | 2903 | 9020525 | ||
BC136536.1 | - | 21 | 2903 | 9020526 |