Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256164.1 | NP_001243093.1 | 7 | 3952 | ||
NM_001256165.1 | NP_001243094.1 | 7 | 4254 | |||
NM_019026.4 | NP_061899.2 | 7 | 4054 | |||
NR_045818.1 | 7 | 3977 |