Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178454.4 | NP_848549.3 | 5 | 471 | ||
XM_005270469.2 | XP_005270526.1 | 6 | 529 | |||
XM_005270470.1 | XP_005270527.1 | 6 | 652 | |||
XM_011540707.1 | XP_011539009.1 | 6 | 470 | |||
XM_011540708.1 | XP_011539010.1 | 6 | 550 | |||
GenBank mRNA | AK309557.1 | - | 5 | 708 | ||
AL833098.1 | - | 5 | 839 | |||
AY336747.1 | - | 5 | 349 | |||
AY358492.1 | - | 5 | 334 | |||
BC047025.1 | - | 4 | 286 | 5491665 | ||
BC091509.1 | - | 6 | 563 | 30513000 | ||
DC339928.1 | - | 5 | 471 | |||
EF710624.1 | - | 3 | 214 |