Search Thermo Fisher Scientific
Gene Symbol: STAT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012448.3 | NP_036580.2 | 16 | 2202 | ||
XM_017024977.1 | XP_016880466.1 | 15 | 2021 | |||
GenBank mRNA | AB208920.1 | - | 16 | 2181 | ||
AK308163.1 | - | 16 | 2090 | |||
BC065227.1 | - | 16 | 2202 | 5553426 | ||
U47686.1 | - | 16 | 2179 | |||
U48730.2 | - | 15 | 2044 |